Learn More
Sklar InstrumentsSupplier Diversity Partner Alm Self-Retaining Retractor, Sklar, 2-3/4 in., 4x4 Prongs Blunt, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 229027
Alm Self-Retaining Retractor, Sklar, 2-3/4 in., 4x4 Prongs Blunt, Stainless Steel. Sklar's Alm Self-Retaining Retractor is an orthopedic instrument commonly used for soft tissue retraction in small bone and joint procedures. It has a thumb screw which opens and locks the arms into place. The arms feature outward curving prongs to hold the tissue securely. This product is straight with 4X4 blunt prongs, and a length of 2-3/4 inches.
Catalog No. 50-143-7925