Learn More
Globe Scientific Self-Standing Transport Tubes with Attached Screw Cap
Designed for the safe storage and transport of biological materials.
$195.10 - $202.50
Specifications
Style Self Standing Transport Tube
Closure Material Polyethylene
Marking Spot Etched
Disposable Yes
Material Polypropylene
View More Specs
Products 2
Catalog Number Mfr. No. Sterility Volume (Metric) Color Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Volume (Metric) Color Price Quantity & Availability  
22-010-1228 View Documents Globe Scientific
6101C
Non-sterile 5 mL Case of 1000 for $195.10
Add to cart
22-010-1233 View Documents Globe Scientific
6102C
Non-sterile 10 mL Case of 1000 for $202.50
Add to cart
Description
Description
  • Tubes are made of polypropylene
  • With molded graduations
  • Self-standing with conical bottom
  • Transparent tubes allows for unobstructed viewing of samples
  • Shatter-proof and may be exposed to freezing temperatures
  • Leak-resistant polyethylene screw caps can be removed or sealed in single turn
Specifications
Specifications
Self Standing Transport Tube
Etched
Polypropylene
Yes (Without Cap)
Polyethylene
Yes
Non-sterile