Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Self Sticking 3-in-1 Sign: CONFINED SPACE ENTER BY PERMIT ONLY

Improve workplace safety with non-adhesive mounted signage. The Brady™ Self Sticking 3-in-1 Self Sticking 3-in-1 Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.

Manufacturer:  Brady™ 102433

 View more versions of this product

Catalog No. 19-107-783


  Check Availability
Add to cart

Specifications

Description

  • This Self Sticking 3-in-1 Sign has diamond-grade reflective properties
  • Strong adhesive back for indoor or outdoor use
Description & Specifications

Specifications

Black/red on glow
7 in.
CONFINED SPACE ENTER BY PERMIT ONLY, Pictogram B
Diamond
10 in.
Not laminated
Vinyl
1/Sign
10 in. x 7 in.
Self-adhesive
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit