Learn More
Brady™ Self-Sticking Pipe Marker for Operations - Pressure Sensitive Vinyl, Style 1, UNBLEACHED STOCK
Self-sticking pipe markers from Brady™ simplify installation with their factory-applied industrial-grade adhesives: simply peel off the backing and press the marker in place.
Manufacturer:  Brady™ 91782
Catalog No. 19-112-176
Add to cart
Description
Description
  • Banding each end of a pipe marker with directional flow arrow tape
  • Style 1HV pipe markers measure 4 in. x 24 in., come with one marker per card and fit 8 in. or greater pipe Dia.
  • High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
  • Good chemical, oil and water resistance
Specifications
Specifications
No
Black on Yellow
Pipe Marker
2.25 in. H x 14 in. W
2.25 in.
Card
2.25H x 14 in. W
14 in.
English
Vinyl
Card
Permanent Acrylic
ANSI A13.1-2007
2.25 in. H x 14 in. W
2 in.
Card
Square
0.010
355.6 mm
Pipe Marker
Pipe Marker