Learn More
Brady™ Self-Sticking Pipe Markers for Other Liquids - Pressure Sensitive Vinyl, Style 4, GLYCOL RETURN
Self-sticking pipe markers from Brady™ simplify installation with their factory-applied industrial-grade adhesives: simply peel off the backing and press the marker in place.
Manufacturer:  Brady™ 94992
Catalog No. 19-112-212
Add to cart
Description
Description
  • Banding each end of a pipe marker with directional flow arrow tape
  • Style 4 pipe markers measure 1-1/8 in. x 7 in., come with four markers per card and fit pipe Dia. 3/4 in. to 2-3/8 in.
  • High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
  • Good chemical, oil and water resistance
Specifications
Specifications
No
Black on Yellow
Pipe Marker
1.125 in. H x 7 in. W
Card of 4 Each
1.125H x 7 in. W
7 in.
English
Vinyl
Card of 4 Each
Permanent Acrylic
ANSI A13.1-2007
1.125 in. H x 7 in. W
1.125 in.
Card of 4 Each
0.010
355.6 mm
Pipe Marker
Pipe Marker