Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SELL Mouse anti-Human, PE, Clone: 4AE56, Abnova™

Mouse monoclonal antibody raised against human SELL.

Manufacturer:  Abnova Corporation MAB12411

Catalog No. 89-934-907


Add to cart

Description

Description

SELL is a cell surface component that is a member of a family of adhesion/homing receptors which play important roles in leukocyte-endothelial cell interactions. The molecule is composed of multiple domains: one homologous to lectins, one to epidermal growth factor, and two to the consensus repeat units found in C3/C4 binding proteins. [provided by RefSeq]

Specifications

Specifications

SELL
4AE56
Mouse monoclonal antibody raised against human SELL.
In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA)
CD62L/LAM-1/LAM1/LECAM1/LNHR/LSEL/LYAM1/Leu-8/Lyam-1/PLNHR/TQ1/hLHRc
Mouse
IgG1
50 Reactions
Store at 4°C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Immunofluorescence
PE
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Liquid
SELL
SELL
Protein A/G affinity chromatography
RUO
Primary
6402
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit