Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SELM Polyclonal Antibody
SELM Polyclonal Antibody
Supplier: Thermo Scientific PA572639
Description
SELM Polyclonal Antibody for Western Blot

Specifications
SELM | |
Polyclonal | |
Unconjugated | |
SELENOM | |
cb693; fc13f12; SELENOM; Selenoprotein M; selenoprotein M precursor; selenoprotein SelM; selm; SelM protein; sepm; sepm protein; wu:fc13f12; zgc:103431 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
352913 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q802G7 | |
SELENOM | |
KLH conjugated synthetic peptide between 96-119 amino acids from the C-terminal region of DANRE selm. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction