Learn More
SELP 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00006403T01
Catalog No. 89-002-046
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
BC068533
6.8
Transient overexpression cell Lysate was tested with Anti-SELP antibody (H00006403-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
SELP 293T Cell Transient Overexpression Lysate(Denatured)
90.8
pCMV-SELP full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.