Learn More
SELP Mouse anti-Human, Clone: Psel.KO.2.5, Abnova™
Mouse monoclonal antibody raised against human SELP.
Manufacturer:  Abnova Corporation MAB12584
Catalog No. 89-934-910
Add to cart
Description
Description
This gene encodes a 140 kDa protein that is stored in the alpha-granules of platelets and Weibel-Palade bodies of endothelial cells. This protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. The membrane protein is a calcium-dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens on neutrophils and monocytes. Alternative splice variants may occur but are not well documented. [provided by RefSeq]
Specifications
Specifications
SELP
Psel.KO.2.5
Mouse monoclonal antibody raised against human SELP.
In PBS, pH 7.4 (0.09% sodium azide).
CD62/CD62P/FLJ45155/GMP140/GRMP/LECAM3/PADGEM/PSEL
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (PFA fixed), Immunoprecipitation
Unconjugated
Flow Cytometry (1:1-1:50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Liquid
SELP
SELP transfected 300.19 cells.
Affinity purification
RUO
Primary
6403