Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SELP Mouse anti-Human, FITC, Clone: 4AW12, Abnova™

Mouse monoclonal antibody raised against human SELP.

Manufacturer:  Abnova Corporation MAB12412

Catalog No. 89-934-908


Add to cart

Description

Description

This gene encodes a 140 kDa protein that is stored in the alpha-granules of platelets and Weibel-Palade bodies of endothelial cells. This protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. The membrane protein is a calcium-dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens on neutrophils and monocytes. Alternative splice variants may occur but are not well documented. [provided by RefSeq]

Specifications

Specifications

SELP
4AW12
Mouse monoclonal antibody raised against human SELP.
In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA)
CD62/CD62P/FLJ45155/GMP140/GRMP/LECAM3/PADGEM/PSEL
Mouse
IgG1
50 Reactions
Store at 4°C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Immunofluorescence
FITC
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Liquid
SELP
SELP
Protein A/G affinity chromatography
RUO
Primary
6403
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit