Learn More
SELP Mouse anti-Human, PE, Clone: 4AW12, Abnova™
Mouse monoclonal antibody raised against human SELP.
Supplier: Abnova Corporation MAB12413
Description
This gene encodes a 140 kDa protein that is stored in the alpha-granules of platelets and Weibel-Palade bodies of endothelial cells. This protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. The membrane protein is a calcium-dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens on neutrophils and monocytes. Alternative splice variants may occur but are not well documented. [provided by RefSeq]
Specifications
SELP | |
Monoclonal | |
PE | |
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user. | |
CD62/CD62P/FLJ45155/GMP140/GRMP/LECAM3/PADGEM/PSEL | |
Mouse | |
Protein A/G affinity chromatography | |
RUO | |
6403 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Flow Cytometry, Immunofluorescence | |
4AW12 | |
Mouse monoclonal antibody raised against human SELP. | |
In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA) | |
SELP | |
SELP | |
50 Reactions | |
Primary | |
Human | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.