Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SELS Mouse anti-Human, Clone: 5G4A10, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261877
Description
SELS Monoclonal antibody specifically detects SELS in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
SELS | |
5G4A10 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
ADO15, MGC104346, SBBI8, selenoprotein S, SelS, SEPS1, VCP-interacting membrane protein, VIMPMGC2553 | |
Mouse | |
IgG1 | |
Protein G purified | |
Signal Transduction | |
Primary | |
55829 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 | |
Purified | |
VIMP | |
Purified recombinant fragment of human VIMP (AA: 1-187) expressed in E. Coli. | |
21.2 kDa | |
0.1 ml | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title