Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01950246
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-246 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-246 Supplier Invitrogen™ Supplier No. PA116932
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In Western Blot, a band is seen at ~50 kDa, representing the secreted form of the protein and also a faint band at ~83 kDa, representing the pro-form of the protein. Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended. Suggested positive control: antigen standard for SEMA3B (transient overexpression lysate), mouse Cerebellum protein, mouse E16 cerebellum lysate.

The semaphorin/collapsin family of molecules plays a critical role in the guidance of growth cones during neuronal development. The secreted protein encoded by this gene family member is important in axonal guidance and has been shown to act as a tumor suppressor by inducing apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA3B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SEMA3B
Gene Accession No. Q13214, Q62177
Gene Alias LUCA-1; Seam V antibody; SemA; sema A; sema A(V); sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B; Sema V; SEMA3B; SEMA5; SEMAA; semaphorin 3B; semaphorin A; semaphorin-3B; semaphorin-A; semaphorin-V; semaV
Gene Symbols SEMA3B
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human SEMA3B protein sequence (between residues 100-200).
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20347, 363142, 484762, 539996, 7869
Target Species Human, Mouse, Rat, Canine, Monkey, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.