Learn More
Aviva Systems Biology Corp SEMA3D Antibody - middle region (ARP55526_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55526_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEMA3D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SEMA3D. Alternate names: MGC39708, Sema-Z2, coll-2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61530