Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEMA4B Polyclonal Antibody
SEMA4B Polyclonal Antibody
Supplier: Thermo Scientific PA533018
Description
SEMA4B Polyclonal Antibody for IHC (P)

Specifications
SEMA4B | |
Polyclonal | |
Unconjugated | |
SEMA4B | |
KIAA1745; mKIAA1745; sema C; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4B; Sema4b; SEMAC; semaphorin 4B; semaphorin-4B; Semaphorin-C; SemC; UNQ749/PRO1480 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10509 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 0.1% sodium azide | |
Q9NPR2 | |
SEMA4B | |
Synthetic 19 amino acid peptide from internal region of human SEMA4B. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction