Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEMA6A Polyclonal Antibody
Supplier: Thermo Scientific PA5111018
Description

Specifications
SEMA6A | |
Polyclonal | |
Unconjugated | |
SEMA6A | |
9330158E07; A730020P05Rik; AI851735; b2b997Clo; HT018; KIAA1368; Mutant line 997; SEMA; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A; sema Q; Sema VIA; SEMA6A; SEMA6A1; SEMA6A-1; semaphorin 6A; semaphorin 6A-1; semaphorin Q; Semaphorin VIA; semaphorin-6A; Semaphorin-6A-1; Semaq; VIA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57556 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2E6 | |
SEMA6A | |
Recombinant Protein corresonding to Human SEMA6A. Recombinant protein control fragment (Product #RP-95589). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction