Learn More
Semaphorin 3C Rat anti-Human, Alexa Fluor 405, Clone: 29U11, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP222041AF405
Catalog No. NB222041405
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Semaphorin 3C Monoclonal antibody specifically detects Semaphorin 3C in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
Semaphorin 3C
29U11
Western Blot, Immunohistochemistry
Purified
SEMA3C
This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human semaphoring 3C. Rat anti human Semaphorin 3C NP_006370.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot, Immunohistochemistry
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
(semaphorin) 3C, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema E, SEMAE, semaphorin E, semaphorin-3C, Semaphorin-E, SemE
Rat
IgG2
0.1 ml
Primary
10512