Learn More
Semaphorin 3C Rat anti-Human, Alexa Fluor™ 532, Clone: 29U11, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222041AF532
Catalog No. NP222041A32
Add to cart
Description
Description
Semaphorin 3C Monoclonal antibody specifically detects Semaphorin 3C in Human samples. It is validated for Western Blot,Immunohistochemistry.
Specifications
Specifications
Semaphorin 3C
29U11
Western Blot, Immunohistochemistry
(semaphorin) 3C, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema E, SEMAE, semaphorin E, semaphorin-3C, Semaphorin-E, SemE
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
SEMA3C
This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human semaphoring 3C. Rat anti human Semaphorin 3C NP_006370.
0.1 mL
Primary
10512