Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 3F Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19148020UL

 View more versions of this product

Catalog No. NBP19148020


Add to cart

Description

Description

Semaphorin 3F Polyclonal antibody specifically detects Antigen in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blotting.
Specifications

Specifications

Semaphorin 3F
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
SEMA3F
Synthetic peptide directed towards the N terminal of human Sema3f. Peptide sequence FIHAELIPDSAERNDDKLYFFFRERSAEAPQNPAVYARIGRICLNDDGGH.
Immunogen affinity purified
RUO
Primary
6405
Western Blot, Immunohistochemistry, Immunoprecipitation
Western Blot 1:1000, Immunohistochemistry, Immunoprecipitation 1:10-1:500
NP_035479
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, 3F, sema III/F, sema IV, SEMA4, SEMA-IV, semaphorin III/F, semaphorin IV
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit