Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 5B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Semaphorin 5B
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095642
View Documents
Novus Biologicals
NBP303805100UL
100 μg Each for $320.71
Add to cart
 
NB095643
View Documents
Novus Biologicals
NBP30380520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Semaphorin 5B Polyclonal antibody specifically detects Semaphorin 5B in Human samples. It is validated for Western Blot
Specifications

Specifications

Semaphorin 5B
Unconjugated
Rabbit
Neuroscience
PBS with 50% glycerol, pH7.3.
54437
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human
FLJ10372, KIAA1445, sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5B
Recombinant fusion protein containing a sequence corresponding to amino acids 800-980 of human Semaphorin 5B (NP_001243276.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit