Learn More
Semaphorin 6B Rat anti-Mouse, Alexa Fluor 405, Clone: RM0317-4N68, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211881AF405
Catalog No. NB211881405
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Semaphorin 6B Monoclonal antibody specifically detects Semaphorin 6B in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Semaphorin 6B
RM0317-4N68
Western Blot
Purified
SEMA6B
Mouse SEMA6B
Protein G purified
Neuroscience
Primary
10501
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B, SEMAN, semaphorin VIB, semaphorin Z, semaphorin-6B, semaphorin-6Ba, SEMA-VIB, semaZ, SEM-SEMA-Y, UNQ1907/PRO4353
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse