Learn More
Semaphorin6C Rat anti-Mouse, Janelia Fluor 646, Clone: RM0318-4K58, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211883JF646
Catalog No. NBP21883646
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Semaphorin6C Monoclonal antibody specifically detects Semaphorin6C in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Semaphorin6C
RM0318-4K58
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
SEMA6C
Mouse SEMA6C
Protein G purified
Neuroscience
Primary
10500
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
KIAA1869, m-SemaY, m-SemaY2, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C, Sema Y, semaphorin-6C, semaphorin-Y, SEMAY
Rat
IgG2
0.1mL
Store at 4°C in the dark.
Monoclonal
Mouse