Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEMG1 Polyclonal Antibody
SEMG1 Polyclonal Antibody
Supplier: Thermo Scientific PA572915
Description
SEMG1 Polyclonal Antibody for Western Blot, IHC (P), IP

Specifications
SEMG1 | |
Polyclonal | |
Unconjugated | |
SEMG1 | |
Alpha-inhibin-31; Alpha-inhibin-92; BB115391; Cancer/testis antigen 103; CT103; dJ172H20.2; MGC14719; RATSVPIIA; semen coagulating protein; semenoclotin; semenogelin I; Semenogelin-1; SEMG; SEMG1; Seminal basic protein; seminal vesicle protein, secretion 1; seminal vesicle protein, secretion 2; seminal vesicle secretory protein 2; seminal vesicle secretory protein II; SGI; SgI-29; Svp1; Svp-1; Svp5; SVPIIA; SVS II; Svs2; SVS2P; Svs2p2; Svs5; SvsII | |
Rabbit | |
50 μL | |
Primary | |
Human, Mouse, Rat, Gerbil | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Conc. Not Determined | |
Whole serum with 0.05% sodium azide | |
P04279 | |
SEMG1, Svs2 | |
Gel purified rat p47 seminal vesicle protein I (secreted Semenogelin I) from rodent seminal vesicle secretion fluid was used as immunogen. | |
RUO | |
25013, 53878, 6406 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction