Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Semiconductor and Chemical Pipe Markers: CARBON DIOXIDE

Identify pipe content and flow direction. Brady™ Semiconductor and Chemical Pipe Markers are ideal for use on small diameter piping or tubing.

Manufacturer:  Brady™ 91907

 View more versions of this product

Catalog No. 19-112-124


  Check Availability
Add to cart

Specifications

Description

  • Meets ASME A13.1 color specifications
  • Made from B-946 self-sticking vinyl with permanent adhesive
  • Use with 1 in. arrow tape
Description & Specifications

Specifications

Acrylic,Cold temperature,Permanent
Legend: CARBON DIOXIDE
W x H: 2.25 x 0.5 in.
Pressure Sensitive Vinyl (B-946)
Green,White
Pipe and Valve Marking
0.004 in.
Semiconductor and Chemical Pipe Marker
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit