Alfa Aesar™ Semiquantitative Standard 1, Specpure™

Manufacturer: Alfa Aesar™ 47180AP

Quantity 500mL

 View more versions of this product

Catalog No. AA47180AP

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD02091742
    UN Number UN3093
    Chemical Name or Material Semiquantitative Standard 1
    Grade Specpure™

    Specifications

    Health Hazard 3 GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD02091742
    UN Number UN3093
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H272-H314-H335
    May intensify fire
    oxidizer.
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.
    Avg. Mol. Wt. or Mol. Wt. Range Matrix: 40% Aqua Regia
    TSCA TSCA
    Physical Form Liquid
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.
    Quantity 500mL
    Chemical Name or Material Semiquantitative Standard 1
    Grade Specpure™

    GHS H Statement
    H272-H314-H335
    May intensify fire
    oxidizer.
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit