Learn More
Semiquantitative Standard 1, Specpure™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 047180AE
Description
Semiquantitative Standard 1, Specpure™ is used as a standard for inductively coupled plasma and direct current plasma (DCP) emission spectrometry for the analysis of trace elements in biomedical and environmental samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Semiquantitative Standard 1 | |
MFCD02091742 | |
Specpure™ | |
Liquid |
100 mL | |
UN3093 | |
Miscible with water |
Safety and Handling
May cause respiratory irritation, May be corrosive to metals, Causes severe skin burns and eye damage
Specific target organ toxicity after single exposure (category 3), Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only