Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sempermed™ SemperGuard™ Powder Free Nitrile Gloves
Click to view available options
Size:
Large
X-Large
Description
- Powder free
- Textured palm and fingers
- Synthetic Nitrile Rubber (SNR)
- Max. 8.0 mil thick at fingertip
- Min. 4.8 mil thick at cuff
- Length: 23 cm
Specifications
Chemotherapeutic Rated | No |
Applications | Laboratory, Industrial |
AQL | 4 |
Color | Blue |
Disposable | Single-use |
Exterior Finish | Partially Textured |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Size | X-Large |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction