Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sempermed™ SemperGuard™ Powdered Nitrile Gloves
SemperGuard™ powdered gloves feature textured fingertips to ensure gripping capabilities in both wet and dry work environments.
Specifications
Applications | Laboratory, Industrial |
---|---|
Certifications/Compliance | FDA 21 CFR Part 177 |
Exterior Finish | Partially Textured |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-818-339
|
Sempermed™
INIPT103 |
Medium | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please Sign In to purchase or view availability for this product. | N/A |
19-818-340
|
Sempermed™
INIPT104 |
Large | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please Sign In to purchase or view availability for this product. | N/A |
Description
- Powdered
- Textured palm and fingers
- Contains rubber latex
- Max. 8.0 mil thick at fingertip
- Min. 4.8 mil thick at cuff
- Length: 23 cm
Specifications
Laboratory, Industrial | |
FDA 21 CFR Part 177 | |
Partially Textured |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title