Learn More
Sempermed™ Best Touch™ Nitrile Powder-Free Gloves
Aloe and vitamin E coating for enhanced protection and comfort
Manufacturer:  Sempermed™ BTNA202
 View more versions of this product
Catalog No. 19-048-611
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Best Touch™ Nitrile gloves with Aloe and Vitamin E is an ideal solution for those who are at risk of becoming sensitive to natural rubber latex. It provides excellent protection and tactile sensitivity while the aloe and vitamin E coating soothes hands and helps prevent irritation of the skin.
  • Chemical resistant gloves manufactured without natural rubber
  • Features Smart Dispense Technology
  • Single Glove Delivery™ packaging method reduces wrinkles and allows for more gloves per box
  • Latex free gloves with operational efficiency
  • Ambidextrous and beaded cuff gloves for single use only
  • Powder-Free gloves eliminate powder aerosolization
  • Rejuvenates and nourishes hands
Certifications
510K approved for medical use
Specifications
Specifications
Safety
Ambidextrous
Nitrile
200 Gloves/Box
Beaded/Rolled
Yes
Small