Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sempermed™ Best Touch™ Nitrile Powder Free Gloves
Click to view available options
Size:
X-Small
Medium
Large
X-Large
Description
- Nitrile gloves with Aloe and Vitamin E prevent skin irritation
- Tactile sensitivity
- Latex-free
- Features Smart Dispense Technology
- Ambidextrous
- Beaded cuffs
- Powder free
Specifications
Applications | Healthcare |
Color | Blue |
Disposable | Single-use |
Exterior Finish | No Texture |
Latex Free | Yes |
Material | Nitrile |
Size | Large |
Thickness (English) | 3.1 mil |
Thickness (Metric) | 0.07 mm |
FDA 510 (k) | Yes |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction