Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sempermed™ Best Touch™ Nitrile Powder Free Gloves

Best Touch™ Nitrile Gloves with Aloe and Vitamin E are ideal for those who are at risk of becoming sensitive to natural rubber latex. It provides protection and tactile sensitivity while the coating soothes hands and helps prevent irritation of the skin.

Specifications

Applications Healthcare
Color Blue
Exterior Finish No Texture
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-048-610
View Documents
Sempermed™
BTNA201
X-Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
19-048-611
View Documents
Sempermed™
BTNA202
Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
19-048-612
View Documents
Sempermed™
BTNA203
Medium This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
19-048-613
View Documents
Sempermed™
BTNA204
Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
19-048-614
View Documents
Sempermed™
BTNA205
X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
Description

Description

  • Nitrile gloves with Aloe and Vitamin E prevent skin irritation
  • Tactile sensitivity
  • Latex-free
  • Features Smart Dispense Technology
  • Ambidextrous
  • Beaded cuffs
  • Powder free
Specifications

Specifications

Healthcare
Blue
No Texture
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit