Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sempermed™ SemperCare™ Tender Touch™ Nitrile Powder Free Gloves

Latex-free exam gloves in a new Smart Dispense Technology, Single Glove Delivery™ packaging. This innovative packaging method reduces wrinkles and allows for more gloves per box.

Manufacturer:  Sempermed™ TTNF203

 View more versions of this product

Catalog No. 19-818-322



Add to cart
This item is not returnable. View return policy

Description

Description

  • Smart Dispense Technology
  • Single glove delivery with fewer wrinkles
  • Synthetic Nitrile Rubber
  • Beaded cuff
  • Fingertip textured
  • Ambidextrous
  • Powder free
  • Tested for use with chemotherapy drugs
Specifications

Specifications

Meets USP 800 requirements for PPE compliance for chemotherapy, FDA 21 CFR part 177
Healthcare
Blue
Partially Textured
Yes
Powder Free
2 mil
Yes
SemperCare™ Tender Touch™
23 cm
Yes
0.65
Yes
No
Nitrile
Medium
0.05 mm
TTNF203
9 in.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit