Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sempermed™ SemperCare™ Tender Touch™ Nitrile Powder Free Gloves

Latex-free exam gloves in a new Smart Dispense Technology, Single Glove Delivery™ packaging. This innovative packaging method reduces wrinkles and allows for more gloves per box.

$78.84 - $119.30

Specifications

Applications Healthcare
Certifications/Compliance Meets USP 800 requirements for PPE compliance for chemotherapy, FDA 21 CFR part 177
Chemotherapeutic Rated Yes
Color Blue
Exterior Finish Partially Textured
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-818-320
View Documents
Sempermed™
TTNF201
X-Small
Pack of 200 for $119.30
Case of 10 PK for $1,136.00
Add to cart
 
19-818-321
View Documents
Sempermed™
TTNF202
Small
Pack of 200 for $119.30
Case of 10 PK for $1,136.00
Add to cart
 
19-818-322
View Documents
Sempermed™
TTNF203
Medium
Pack of 200 for $119.30
Case of 10 PK for $1,136.00
Add to cart
 
19-818-323
View Documents
Sempermed™
TTNF204
Large
Pack of 200 for $119.30
Case of 10 PK for $1,136.00
Add to cart
 
19-818-324
View Documents
Sempermed™
TTNF205
X-Large
Pack of 180 for $78.84
Case of 10 PK for $788.40
Add to cart
 
Description

Description

  • Smart Dispense Technology
  • Single glove delivery with fewer wrinkles
  • Synthetic Nitrile Rubber
  • Beaded cuff
  • Fingertip textured
  • Ambidextrous
  • Powder free
  • Tested for use with chemotherapy drugs
Specifications

Specifications

Healthcare
Yes
Partially Textured
Meets USP 800 requirements for PPE compliance for chemotherapy, FDA 21 CFR part 177
Blue
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit