Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sempermed™ SemperSure™ Powder Free Nitrile Gloves

SemperSure™ Nitrile Exam Gloves are designed for those with skin sensitivities to regular nitrile gloves. Accelerator-free gloves offer all the benefits of a nitrile glove, minus the sulfur-based chemical accelerators that cause Type I (latex) and Type IV (chemical) allergies.

$89.85 - $138.70

Specifications

Applications Healthcare, Laboratory
Certifications/Compliance Meets 800 USP requirements for PPE compliance for chemotherapy, FDA 21 CFR Part 177
Chemotherapeutic Rated Yes
Color Blue
Exterior Finish Partially Textured
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-818-325A
View Documents
Sempermed™
SUNF201
X-Small
Pack of 200 for $138.70
Case of 10 PK for $1,321.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-818-325B
View Documents
Sempermed™
SUNF202
Small
Pack of 200 for $138.70
Case of 10 PK for $1,321.00
Add to cart
 
19-818-325C
View Documents
Sempermed™
SUNF203
Medium
Pack of 200 for $138.70
Case of 10 PK for $1,321.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-818-325D
View Documents
Sempermed™
SUNF204
Large
Pack of 200 for $138.70
Case of 10 PK for $1,321.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-818-325E
View Documents
Sempermed™
SUNF205
X-Large
Pack of 180 for $89.85
Case of 10 PK for $898.50
Add to cart
 
Description

Description

  • Synthetic Nitrile Rubber
  • Accelerator-free
  • Low dermatitis potential
  • Fingertip textured
  • Ambidextrous
  • Powder free
  • Tested for use with chemotherapy drugs
Specifications

Specifications

Healthcare, Laboratory
Yes
Partially Textured
Meets 800 USP requirements for PPE compliance for chemotherapy, FDA 21 CFR Part 177
Blue
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit