Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sempermed™ StarMed™ Latex Gloves

Powder free latex exam gloves are designed to provide protection and tactile sensitivity for healthcare and laboratory applications.

Specifications

Applications Healthcare, Laboratory
Thickness (English) 5.5 mil
Thickness (Metric) 0.13 mm
Exterior Finish Fully Textured
Length (English) 9 in.
View More Specs

Alert:

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-123-554
View Documents
Sempermed™
SM105
X-Large
Pack for $50.30
Case of 10 PK for $479.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-123-550
View Documents
Sempermed™
SM101
X-Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
19-123-551
View Documents
Sempermed™
SM102
Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
19-123-552
View Documents
Sempermed™
SM103
Medium This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
19-123-553
View Documents
Sempermed™
SM104
Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
Description

Description

  • Advanced technology online chlorination process
  • Reduced tackiness, quick and easy to don
  • 50 micrograms or less of total water-extractable proteins per gram
  • Micro-rough textured surface
  • Tactile sensitivity with increased gripping characteristics
Specifications

Specifications

Healthcare, Laboratory
0.13 mm
9 in.
5.5 mil
4.6 mil
5.5 mil
Fully Textured
22.8 cm
0.13 mm
0.11 mm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit