Learn More
Tocris Bioscience SEN 1269
Amyloid-β aggregation inhibitor
Manufacturer: Tocris Bioscience 4699/50
This item is not returnable.
View return policy
Description
Amyloid-β (Aβ) aggregation inhibitor; blocks Aβ(1-42) aggregation and protects neuronal cell lines exposed to Aβ(1-42). Reduces Aβ oligomer-induced LTP and memory deficits in vivo.Specifications
SEN 1269 | |
>98 | |
50mg |
SEN 1269 | |
322.36 |