Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SENP2 Mouse anti-Human, Non-human primate, Clone: OTI3E7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526377

Catalog No. PIMA526377


Add to cart

Description

Description

SUMO (small ubiquitin-related modifier), a member of the ubiquitin-like protein family, regulates diverse cellular functions of a variety of target proteins including transcription, DNA repair, nucleocytoplasmic trafficking and chromosome segregation. SUMO precursor proteins undergo cleavage of the residues after the ""GG"" region by SUMO-specific proteases in maturation. This cleavage of the precursor is a prerequisite for subsequent sumoylation. The sentrin-specific (or SUMO-specific) protease (SENP) proteins belong to the peptidase C48 family and include SENP1-3 and SENP5-8. SENP1, SENP2 and SENP3 degrade UBL1 and SMT3H2 conjugates and subsequently release the monomers from sumoylated substrates.
Specifications

Specifications

SENP2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HC62
AXAM2, SMT3IP2, SMT3-specific isopeptidase 2, SUMO1/sentrin/SMT3 specific protease 2, sentrin-specific protease 2, sentrin/SUMO-specific protease SENP2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
59343, 700717
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3E7
Unconjugated
SENP2
Liquid
SENP2
Human recombit protein fragment corresponding to amino acids 139-523 of SENP2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Non-human primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit