Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SENP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15659020UL
Description
SENP3 Polyclonal specifically detects SENP3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.Specifications
SENP3 | |
Polyclonal | |
Western Blot 0.2-1 ug/ml, Immunohistochemistry, Immunofluorescence 1:10-1:500 | |
DKFZP586K0919, DKFZp762A152, EC 3.4.22, EC 3.4.22.-, sentrin/SUMO-specific protease 3, Sentrin/SUMO-specific protease SENP3, sentrin-specific protease 3, SMT3IP1, SSP3DKFZp586K0919, SUMO1/sentrin/SMT3 specific peptidase 3, SUMO1/sentrin/SMT3 specific protease 3, SUMO-1-specific protease 3, SUSP3 | |
Rabbit | |
Affinity Purified | |
RUO | |
26168 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
SENP3 | |
Synthetic peptides corresponding to SENP3(SUMO1/sentrin/SMT3 specific peptidase 3) The peptide sequence was selected from the N terminal of SENP3. Peptide sequence PPPKPRLKSGGGFGPDPGSGTTVPARRLPVPRPSFDASASEEEEEEEEEE. | |
20 μL | |
Primary | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction