Learn More
Aviva Systems Biology Corp SENP6 Antibody - C-terminal region (ARP50600_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50600_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SENP6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SENP6. Alternate names: FLJ11355, FLJ11887, KIAA0389, KIAA0797, SSP1, SUSP1ApplicationWB. Species reactivity: Cow, Horse, Human, Pig
Catalog No. 50-014-60385