Learn More
Sensidyne™ Colorimetric Gas Detector Tubes: 2-Butanol
Manufacturer:  Sensidyne™ 189U
Catalog No. 18-073J
Add to cart
Specifications
Specifications
2-Butanol
4 to 300ppm
10 tubes/box
Standard tube