Learn More
Sensidyne™ Colorimetric Gas Detector Tubes: Cyclohexanol
Supplier: Sensidyne™ 206U
Specifications
Cyclohexanol | |
10 Tubes/Box | |
10° to 40°C |
5 to 500ppm | |
50°F to 104°F | |
Standard tube |