Learn More
Sensidyne™ Colorimetric Gas Detector Tubes: Isopentyl Acetate
Manufacturer:  Sensidyne™ 188U
Catalog No. 18-070W
Add to cart
Specifications
Specifications
Isopentyl Acetate
10 tubes/box
10° to 40°C
10 to 400ppm
50° to 104°F
Standard tube