Learn More
Sensidyne™ Colorimetric Gas Detector Tubes: Methyl Cyclohexanol
Supplier: Sensidyne™ 199U
Specifications
Methyl Cyclohexanol | |
10 Tubes/Box | |
10° to 40°C |
5 to 200ppm | |
50°F to 104°F | |
Standard tube |