Learn More
Sensidyne™ Colorimetric Gas Detector Tubes: N,N-Dimethylformamide
Supplier: Sensidyne™ 196S
Specifications
N,N-Dimethylformamide | |
10 Tubes/Box | |
10° to 40°C |
1 to 30ppm | |
50°F to 104°F | |
Standard tube |