Learn More
Sensidyne™ Detector Tubes: Phosphine
Supplier: Sensidyne™ 121SD
Description
All detector tubes are sold in packs of 10.
Specifications
Phosphine | |
0.5 to 1 | |
1 min. | |
0°C to 40°C |
0.25 to 20ppm | |
10 | |
32°F to 104°F |