Learn More
Sensidyne™ Detector Tubes: Sulfur Compounds
Supplier: Sensidyne™ 103SA
Description
All detector tubes are sold in packs of 10.
Specifications
Sulfur Dioxide | |
1 | |
1 min. | |
0°C to 40°C |
0.1 to 3% | |
10 | |
32°F to 104°F |