Learn More
Thermo Scientific™ Sensititre™ SWIN™ Software System Available on GSA/VA Contract for Federal Government customers only.
Consolidate your entire test program on a single software platform while enhancing data entry and reporting with the Thermo Scientific™ Sensititre™ SWIN™ Software System.
Manufacturer:  Thermo Scientific™ SW4000
 View more versions of this product
Catalog No. STSW4000
Add to cart
Description
Description
Consolidate your entire test program on a single software platform while enhancing data entry and reporting with the Thermo Scientific™ Sensititre™ SWIN™ Software System. Simple touch screen navigation puts results at your fingertips while drop-down fields eliminate manual entry errors for all manual, semi-automated and fully automated reads.
  • Customizable Expert System provides three-tiered expert messages: Information, Modification (based on FDA, CLSI, and EUCAST recommendations) and Warning Levels
  • Quality control module to easily manage MIC results and broth information, for inspection purposes
  • Automated alerts and report functionality enhance workflow and expedite release of results
  • View ID details and susceptibility results at the same time
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Sensititre SWIN Software, Complete