Learn More
Vileda Professional Sentrex Cut End String Mop
Ideal for rugged cleaning on rough surfaces and high traffic areas
Manufacturer: Vileda Professional 134837
Description
- Made from highly durable synthetic/rayon blended yarn with superior tensile strength properties
- Can be used for floor finishing or waxing
- Delivers high absorbency and can withstand repeated laundering without linting
- Heavy duty poly-coated mesh headband withstands harsh chemicals and launderings
Specifications
12/Cs. | |
White | |
20 oz. | |
Synthetic/Rayon Blended Yarn | |
String Mop |
Not Autoclavable | |
Cut end, narrow band | |
Rough surfaces, high traffic areas | |
Non-sterile |