Learn More
Alfa Aesar™ Sepabeads™ SP710, synthetic adsorbent resin, highly porous type, PS-DVB, P.R. 90 angstroms
Manufacturer:  Alfa Aesar™ 46555A1
Quantity 1kg
 View more versions of this product
Catalog No. AA46555A1
  Check Availability
Add to cart
Specifications
Description
  • Synthetic adsorbents for juice debittering and decolorizing
  • Also highest naringin capacity
Description & Specifications
Chemical Identifiers
Sepabeads™ SP710, synthetic adsorbent resin, Highly porous type, PS-DVB
Specifications
250μm, P.R. 90Å
1.01g/mL
900m2/g
Beads
1kg
Sepabeads™ SP710, synthetic adsorbent resin, Highly porous type, PS-DVB