Learn More
GVS™ SEPARA™ 32 mm Syringeless Filter Vial
The SEPARA™ single step filtering process is efficient and simple to use
$181.10 - $196.10
Specifications
Capacity 450 μL
Diameter (Metric) 12 mm
Housing Material PE
Includes Sealed membrane chamber and cap with a pre-cut septa.
Length (English) 33 mm
View More Specs
Products 10
Catalog Number Mfr. No. Pore Size Color Membrane Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size Color Membrane Price Quantity & Availability  
10-486-204
View Documents
GVS™
MV32ANPNY002BC01
0.2 μm Blue Nylon (NY) Pack of 100 for $192.10
Add to cart
10-486-205
View Documents
GVS™
MV32ANPNY004UC01
0.45 μm Blue Nylon (NY) Pack of 100 for $181.10
Add to cart
10-486-208
View Documents
GVS™
MV32ANPPS002EC01
0.2 μm Green Polyethersulfone (PES) Pack of 100 for $181.10
Add to cart
10-486-209
View Documents
GVS™
MV32ANPPS004WC01
0.45 μm Green Polyethersulfone (PES) Pack of 100 for $181.10
Add to cart
10-486-200
View Documents
GVS™
MV32ANPPT002TC01
0.2 μm Pink Polytetrafluoroethylene (PTFE) Pack of 100 for $196.10
Add to cart
10-486-201
View Documents
GVS™
MV32ANPPT004CC01
0.45 μm Red Polytetrafluoroethylene (PTFE) Pack of 100 for $196.10
Add to cart
10-486-206
View Documents
GVS™
MV32ANPPV002FC01
0.2 μm Yellow Polyvinylidene Fluoride (PVDF) Pack of 100 for $192.10
Add to cart
10-486-207
View Documents
GVS™
MV32ANPPV004IC01
0.45 μm Orange Polyvinylidene Fluoride (PVDF) Pack of 100 for $181.10
Add to cart
10-486-202
View Documents
GVS™
MV32ANPRC002GC01
0.2 μm Gray Regenerated Cellulose (RC) Pack of 100 for $196.10
Add to cart
10-486-203
View Documents
GVS™
MV32ANPRC004LC01
0.45 μm Black Regenerated Cellulose (RC) Pack of 100 for $196.10
Add to cart
Description
Description
  • Rapid sample preparation.
  • Filtering with a plunger in the vial is a rapid single step, reducing sample loss.
  • After filtration, the sample is ready for use in auto-sampling robot.
  • The pre-slit cap ensures easy and clean transfer of sample.
  • Compatible with most standard auto-Samplers
Applications by filtration membrane:
PTFE
  • Filtration of strong acids and aggressive solutions
Nylon
  • Clarification of aqueous and organic solvent solutions
  • HPLC sample preparation
  • Chromatography
Membrane RC
  • Filtration of Aqueous and Organic Solutions
  • Analytical Sample Prep, uHPLC
  • Chromatography
  • Clarification
  • Protein Chemistry
Specifications
Specifications
450 μL
PE
33 mm
Non-sterile
50°C
480 μL
12 mm
Sealed membrane chamber and cap with a pre-cut septa.
100
120°F
Syringeless